Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Misoprostol, Cytoprotective PGE2 Analog, Liquid, 10 mg

Catalog No. 502707148
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7148 Supplier Abcam Supplier No. AB12090210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytoprotective PGE2 analog with activity mediated by EP2, EP3, and EP4 receptors.

  • Molecular Weight: 382.54 g/mol
  • Purity: >98%
  • Description: Cytoprotective PGE2 analog with activity mediated by EP2, EP3, and EP4 receptors (Ki values are 250, 67 and 67 nM, respectively)
  • Form: Liquid
  • Storage Buffer: 99% Methyl acetate, 1% Misoprostol
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Blue Ice
  • CAS Number: 59122-46-2

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Liquid
Shipping Condition Blue Ice
Content And Storage Store at -20°C.
Substrate Properties Chemical Substrate
Target Molecule EP2, EP3, EP4 Receptors
Formulation 99% Methyl acetate, 1% Misoprostol
Quantity 10 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.