Learn More
Mitchell Plastics Biohazard Benchtop Waste Containers
Description
- Made of heavy-duty acrylic to help protect personnel from exposure to potential hazards
- Clear design enables warning insignias and biohazard bag colors to be seen
- Each unit has a hinged lid for easy placement of waste
- Lids are also removable for easy positioning and removal of bag or box
- Reusable
Bench top models
- WC-300: holds bag 35.56 × 48.26cm (14 × 19 in.)
- WC-400: holds bag 20.32 × 30.48cm (8 × 12 in.)
- WB-600: holds glass disposal box 20.32 × 20.32 × 25.4cm (8 × 8 × 10 in.)
Specifications
Specifications
| Color | Clear |
| Dimensions (L x W x H) | 4 x 4.5 x 8.5 in. (10.16 x 11.43 x 21.6 cm) |
| Height (Metric) | 21.6 cm |
| Length (Metric) | 10.16 cm |
| Model | Benchtop |
| Size | Medium |
| Width (Metric) | 11.43 cm |
| For Use With (Application) | Waste disposal |
| For Use With (Equipment) | 20.32 x 30.48 cm Bag |
| Width (English) | 4.5 in. |
| Show More |
Dispose contents according to all local, state and federal disposal regulations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.