Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mitchison 7H11 Selective Agar with Carbenicillin, Polymyxin B, Amphotericin B and Trimethoprim

Catalog No. R08576
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08576 20/Pk. F - slant
R01610 10/Pk. Monoplate Deep Pour
R08574 100/Pk. F - slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R08576 Supplier Thermo Scientific™ Supplier No. R08576
This item is not returnable. View return policy

Isolate and cultivate Mycobacterium species from specimen containing mixed flora with this selective medium.

Isolate and cultivate Mycobacterium species from specimen containing mixed flora with Thermo Scientific™ Remel™ Mitchison 7H11 Selective Agar with Carbenicillin, Polymyxin B, Amphotericin B, and Trimethoprim. The presence of casein hydrolysate in the medium enhances growth of Mycobacterium species more than on Middlebrook 7H10. Different antibiotics act as selective agents against most Gram-positive and Gram-negative bacteria.

Contains carbenicillin, polymyxin B, amphotericin B, trimethoprim
TRUSTED_SUSTAINABILITY

Specifications

Description Mitchison 7H11 Selective Agar w/carbenicillin, polymyxin B, amphotericin B, trimethoprim
Format F - slant
Product Type Agar Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.