Learn More
VAI Mitcool 1700 MP Cleanroom Lab Coats
Catalog No. 19546405 Shop All VAI Products
Change view
Click to view available options
Size:
Small
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-546-405 -
19-547-247 Small
19-546-403 -
19-546-404 -
19-546-406 -
19-547-252 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-546-405 Supplier VAI Supplier No. 1700LCS1723
Add to Cart
Add to Cart
This item is not returnable. View return policy
MP lab coats are constructed of a non-woven, microporous, coated material and provide protection in controlled environments.
  • Bonded polypropylene with laminate
  • Liquid chemical resistance
  • Comfortable, yet durable
  • Low particle shedding
  • Disposable
Specifications
Product Type Controlled Environments Lab Coat
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Seam Type Bound
Certifications/Compliance ASTM F903-96, ASTM F1407-96
Construction Polypropylene, Laminate Binding Material