Learn More
Biocare Medical, LLC Microphthalmia Transcription Factor (MiTF) Antibody - Concentrate, 0.5 ml
SDP
Supplier:  Biocare Medical, LLC CM423BK
Encompass_Preferred
Microphthalmia Transcription Factor antibody (MiTF) is a nuclear melanocytic marker. Studies have shown it is a sensitive and specific marker for malignant melanoma, including some spindle-cell variants. MiTF has been shown to have superior sensitivity and specificity to S100 and HMB45. MiTF may be useful for identification of melanoma, melanocytic soft tissue tumors and the unusual group of tumors that show combined melanocytic and myloid differentiation, the perivascular epithelioid cell family of tumors (PEComas). Microphthalmia Transcription Factor may be a valuable addition to a melanoma marker panel with S-100, HMB45, Tyrosinase and MART-1.
Catalog No. 50-827-15
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.