Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MiTF Recombinant Rabbit Monoclonal Antibody (JF100-01)
GREENER_CHOICE

Catalog No. PIMA532554 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532554 Supplier Invitrogen™ Supplier No. MA532554
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, while the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MiTF
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone JF100-01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene MITF
Gene Accession No. O75030, O88368, Q08874
Gene Alias BCC2; Bhlhe32; black eyed white; bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; mi; microphtalmia-associated transcription factor; microphthalmia; microphthalmia associated transcription factor isoform M-; microphthalmia transcription factor; microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; MITF-M-; transcription factor, isoform 7; vit; Vitiligo; Wh; WS2; WS2A
Gene Symbols MITF
Host Species Rabbit
Immunogen Recombinant protein within Human MiTF aa 353-520.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17342, 25094, 4286
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.