Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane, 96%, ACS reagent

Catalog No. AC424010010
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -29°C
Density 1.1200g/mL
Boiling Point 100°C to 102°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.3805 to 1.3825
Linear Formula CH3NO2
Quantity 1 L
Beilstein 01, 74
Fieser 01,739; 04,357; 09,323; 13,199
Specific Gravity 1.12
Merck Index 15, 6698
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
Viscosity 0.61 mPa.s (25°C)
Formula Weight 61.04
Percent Purity 96%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Nitromethane, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh
EINECSNumber 200-876-6
RTECSNumber PA9800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.