Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Mithramycin A, 90%, from Streptomyces argillaceus

Catalog No. AC415090050
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415090050 5 mg Glass Bottle
AC415090010 1 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415090050 Supplier Thermo Scientific Chemicals Supplier No. 415090050
  • $614.50
    $522.65 / Each of 1

    Save $91.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24
Molecular Weight (g/mol) 1085.156
MDL Number MFCD00135618
InChI Key CFCUWKMKBJTWLW-ZUZMCERRSA-N
PubChem CID 122172942
IUPAC Name (2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dih
SMILES CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)O

Specifications

Type Mithramycin A
Physical Form Powder
CAS Min % 88.0
Melting Point 180.0°C to 183.0°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Brown to Red
Quantity 5 mg
Assay Percent Range 90%
Merck Index 15, 7652
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in lower alcohols, acetone, ethyl acetate, moderatly soluble in chloroform, slightly soluble in ether, benzene
Formula Weight 1085.16
Percent Purity 90%
Chemical Name or Material Mithramycin A, 90%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Harmful if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber PZ2800000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.