accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(S)(S)O(S)(R)O(S)(R)HO(S)OH(R)CH3O(R)OH(R)CH3OCH3OHOHO(S)O(S)(R)O(S)(R)O(S)(S)CH3OH(R)OH(R)H3CO(R)OH(R)H3CO(R)OH(R)H3COO(S)OH(R)H3COH

CAS RN 18378-89-7

IUPAC Name: (2S,3S)-3-[(1S,3S,4R)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-2-{[(2S,4R,5R,6R)-4-{[(2S,4R,5R,6R)-4-{[(2S,4S,5R,6R)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-6-{[(2S,4R,5R,6R)-4-{[(2S,4R,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-8,9-dihydroxy-7-methyl-1,2,3,4-tetrahydroanthracen-1-one
Synonyms: Mith antibiotic LA 7017 mitramycin mitramycine MTH mitramycinum plicamycin preparation plicamycin (substance) plicamicina aurlelic acid
Molecular Weight (g/mol): 1085.16
Molecular Formula: C52H76O24
InChi Key: CFCUWKMKBJTWLW-WZIVUMCRSA-N
SMILES: CO[C@@H]([C@@H]1CC2=C(C(=O)[C@H]1O[C@H]1C[C@@H](O[C@H]3C[C@@H](O[C@H]4C[C@](C)(O)[C@H](O)[C@@H](C)O4)[C@H](O)[C@@H](C)O3)[C@H](O)[C@@H](C)O1)C(O)=C1C(O)=C(C)C(O[C@H]3C[C@@H](O[C@H]4C[C@@H](O)[C@H](O)[C@@H](C)O4)[C@H](O)[C@@H](C)O3)=CC1=C2)C(=O)[C@@H](O)[C@@H](C)O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)