Learn More
Apexbio Technology LLC Mitiglinide Calcium 145525-41-3 1g
SDP
Supplier:  Apexbio Technology LLC B21931000
Encompass_Preferred
Mitiglinide Calcium (CAS 145525-41-3) is a small-molecule antagonist selectively targeting ATP-sensitive potassium (KATP) channels By binding and inhibiting these KATP channels in pancreatic beta cells mitiglinide calcium induces membrane depolarization and prompts calcium influx thus stimulating insulin secretion This mechanism positions the molecule as relevant for biomedical research investigating glucose regulation pathways beta cell function and pathophysiology of type 2 diabetes mellitus
Catalog No. 50-000-62606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.