Learn More
Sartorius Mito Membrane Potential
Supplier:  Sartorius 91038
Encompass
No wash protocol – just add and read for fast easily automated workflow.Multiplex with Qbeads PlexScreen kits for richer content.PrincipleMitochondrial membrane potential is determined by sequestration of a small fluorescent molecule inside the lumen of intact mitochondria with an active membrane potential. Upon mitochondrial depolarization the dye leaks into the cytoplasm and the cell exhibits a decrease in fluorescence.
Catalog No. 50-289-1598