Learn More
Agilent Technologies Mito-rOCR Assay
Catalog No. MO4004
Change view
Click to view available options
Includes:
Magnetic holder only
Mito-rOCR Analysis Module software, 3 license seats
Mito-rOCR microplates and seal lids only
Reagents, plates and seal lids for 4 assays
Starter kit: reagents, plates, seal lids, magnetic holder and software (3 license seats)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
MO4004 Starter kit: reagents, plates, seal lids, magnetic holder and software (3 license seats)
MO3004 Reagents, plates and seal lids for 4 assays
MO100 Magnetic holder only
MO2004 Mito-rOCR microplates and seal lids only
MO500 Mito-rOCR Analysis Module software, 3 license seats
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MO4004 Supplier Agilent Technologies Supplier No. MO4004
Add to Cart
Add to Cart
Request a Quote
The Agilent Mito-rOCR Assay makes mitochondrial function analysis easy and accessible. Measure relative oxygen consumption rate on a plate reader.
  • Simplifies mitochondrial respiration assessment with an intuitive, easy-to-use design.
  • Provides quick & reliable rOCR measurements for efficient data collection.
  • Eliminates the need for oil with Mito-rOCR seal lids, streamlining the assay process.
  • Compatible with a wide range of BioTek fluorescence plate readers for versatile use.
  • Glass-like plate bottom allows for data multiplexing and imaging of the same samples post-assay.
  • Automated data analysis module accelerates research with fast and robust results.
  • Reduces assay and analysis time by up to five times compared to traditional oil-based methods.
Specifications
Detection Method Fluorescence
For Use With (Equipment) Synergy H1 Hybrid Multi-Mode Reader, Cytation 1 Cell Imaging Multi-Mode Reader, Cytation 5 Cell Imaging Multi-Mode Reader, Cytation 7 Cell Imaging Multi-Mode Reader, Cytation C10 Confocal Imaging Reader, Synergy Neo2 Hybrid Multi-Mode Microplate Reader
Includes Starter kit: reagents, plates, seal lids, magnetic holder and software (3 license seats)
Quantity 4 Tests
Type Cell-Based Assay Kit
Cell Line Any Adherent Cell Type
System Type Mito-rOCR
Druggable Target Cell-based Assay