Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MitoBrilliant 646
GREENER_CHOICESDP

Catalog No. 77001PACK
Change view
Click to view available options
Quantity:
1 Pk. (5 x 50 μg)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77001PACK 1 Pk. (5 x 50 μg)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77001PACK Supplier Tocris Bioscience™ Supplier No. 7700/1 PACK

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Universal red fluorescent mitochondrial stain for both live and fixed cells

MitoBrilliant probes have been developed to overcome some common limitations encountered with standard mitochondrial trackers, offering clearer answers to scientific questions. They are next-generation fluorescent stains for the localization and tracking of mitochondria in both live and fixed cells. The range harnesses Janelia Fluor® dye technology, conferring some of the properties that make these widely used dyes, into mitochondrial stains.

MitoBrilliant™ 646 is a next-generation universal stain for mitochondria in both live and fixed cells. It is retained in mitochondria following fixation with exceptionally clear staining and is retained in the mitochondria of live cells following loss of the mitochondrial membrane potential. Additionally, MitoBrilliant™ 646 can be used to stain pre-fixed cells/tissue, making this dye suitable for IHC/ICC.

Two dyes that accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner are also available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).

Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.

MitoBrilliant is a trademark of Bio-Techne Corp.

Specifications

Content And Storage Store at -20°C
Quantity 1 Pk. (5 x 50 μg)
Target Mitochondria Dyes and Probes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.