Learn More
Abcam Mitochondrial Hydroxyl Radical Detection Assay Kit, Fluorescent, 200 Test
Description
- Detection method: Fluorescent
- Sample types: Suspension cells, Adherent cells
- Compatible instruments: Microplate reader, Fluorescence microscope
- Cited in: > 5 publications (7 total)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Hydroxyl Radical |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Microplate Reader, Fluorescence Microscope |
| Format | Kit |
| No. of Reactions | 200 Tests |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 200 Tests |
| For Use With (Application) | Hydroxyl radical detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.