Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondrial Pyruvate Carrier 2 (BRP44), Mouse anti-Human, Mouse, Clone: , Millipore Sigma™

Catalog No. MABS191425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS191425 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS191425 Supplier MilliporeSigma Supplier No. MABS191425UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mitochondrial Pyruvate Carrier 2 (BRP44), Cat. No. MABS1914, is a mouse monoclonal antibody that detects human and mouse Mitochondrial pyruvate carrier 2 and has been tested for use in Immunoprecipitation and Western Blotting.

Mitochondrial pyruvate carrier 2 (UniProt: O95563; also known as Brain protein 44, MPC2) is encoded by the MPC2 (also known as BRP44) in human. MPC2 is a multi-pass inner mitochondrial membrane protein that mediates the imports pyruvate into mitochondria. Pyruvate is an essential component in intermediary metabolism that occupies a key crossroads position between cytosolic glycolysis and mitochondrial metabolism. MPC2 contains 3 helical domains (aa 41-61; 73-90; and 96-115). In mammalian cells mitochondrial pyruvate carrier is composed on two paralogous subunits known as MPC1 and MPC2 that heteromerize to form a functional complex. It has been reported that MPC2 can also homomerize, however only MPC1/MPC2 heteromeric forms are functional. Complete absence of MPC2 is shown to cause embryonic lethality in murine models, however, murine cell lines that express an N-terminal truncated MPC2 protein (Mpc2 16) are reported to be viable, but exhibit significantly reduced capacity for mitochondrial pyruvate oxidation. (Ref.: Compan, V., et al. (2015). Mol. Cell 59(3); 491-501; Vigueira, PA., et al. (2014). Cell Rep. 7(6); 2042-2053).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mitochondrial Pyruvate Carrier 2 (BRP44)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: 1μg/mL from a representative lot detected Mitochondrial Pyruvate Carrier 2 (BRP44) in mouse heart and mouse liver tissue lysates. Immunoprecipitation Analysis: A 1:1,000 dilution from a representative lot immunoprecipitated Mitochondrial Pyruvate Carrier 2 (BRP44) in mouse liver mitoplasts (Courtesy of Prof. Jean-Claude Martinou, Ph.D., University of Geneva, Switzerland).. Western Blotting Analysis: 4 μg/mL from a represenetative lot detected Mitochondrial Pyruvate Carrier 2 (BRP44) in mouse pancreas tissue lysate. Western Blotting Analysis: A representative lot detected Mitochondrial Pyruvate Carrier 2 (BRP44) in Western Blotting applications (Compan, V., et. al. (2015). Mol Cell. 59(3):491-501; Vanderperre, B., et. al. (2016). PLoS Genet. 12(5):e1006056).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Mitochondrial pyruvate carrier 2;Brain protein 44;BRP44
Gene Symbols MCP2;BRP44
Host Species Mouse
Immunogen A linear peptide corresponding to 10 amino acids from the internal region of human mitochondrial pyruvate carrier 2 protein.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This mouse monoclonal antibody detects Mitochondrial Pyruvate carrrier Protein 2 in human and mouse cells. It targets an epitope within 10 amino acids from the internal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.