Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitoferrin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735350 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5350 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5350 Supplier Proteintech Group Inc Supplier No. 264691AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.

Specifications

Antigen Mitoferrin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SLC25A37
Gene Accession No. Q920G8, Q9NYZ2
Gene Alias MFRN, Mfrn1, Mitochondrial iron transporter 1, Mitoferrin 1, SLC25A37
Gene Symbols SLC25A37
Host Species Rabbit
Immunogen Mitoferrin 1 Fusion Protein Ag24876
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51312, 67712
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.