Learn More
Tocris Bioscience™ MitoMark Green I

Description
It is recommended to prepare stock solutions in DMSO.
Specifications
Specifications
| Chemical Name or Material | 2-[3-[5,6-Dichloro-1,3-bis[[4-(chloromethyl)phenyl]methyl]-1,3-dihydro-2 H-benzimidazol-2-ylidene]-1-propen-1-yl]-3-methylbenzoxazolium chloride |
| CAS | 201860-17-5 |
| Quantity | 1 Pk. (10 x 50 μg) |
| Target | Mitochondria Dyes and Probes |
| Molecular Formula | C34H28Cl5N3O |
| Formula Weight | 671.87 |
| Purity | 85% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.