Learn More
Enzo Life Sciences Mitomycin C (10mg). CAS: 50-07-7
Supplier: Enzo Life Sciences BMLGR3110010
Antitumor antibiotic. DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway. Purity: ≥95% (HPLC). Solubility: Soluble in water (1mg/ml, warm), 100% ethanol (1mg/ml, warm), DMSO (15mg/ml) or methanol. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.