Learn More
Lkt Laboratories CAS 50-07-7, ≥98%, DNA cross-linker and thioredoxin reductase inhibitor used to treat various cancers.
SDP
Supplier:  Lkt Laboratories M3377
Encompass_Preferred
Mitomycin C is an alkaloid initially produced by Streptomyces that exhibits antibiotic, immunomodulatory, and anticancer chemotherapeutic activities. Mitomycin C increases heart allograft transplant survival rates by decreasing activation of CD4+ T cells and lymph nodes and increasing the number of circulating Treg cells. Mitomycin C inhibits thioredoxin reductase and induces DNA cross-linking at CpG sequences. Clinically, mitomycin C is used as a combination therapy to treat cancers; it suppresses cell growth by preventing DNA strand break repair. 
Catalog No. NC2539786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.