Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mitomycin C, Anticancer and Antibiotic Agent, Solid, 10 mg

Catalog No. 502584569
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4569 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4569 Supplier Abcam Supplier No. AB12079710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent anticancer and antibiotic agent that crosslinks DNA to inhibit synthesis and induce apoptosis.

  • Molecular Weight: 334.33 Da
  • Purity: >99%
  • Physical State: Solid
  • Function: Anticancer and antibiotic agent; crosslinks DNA; inhibits DNA synthesis and nuclear division; induces apoptosis
  • CAS Number: 50-07-7
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule DNA
Buffer 99% Mitomycin C
Content And Storage Store at 4°C.
Form Solid
Quantity 10 mg
For Use With (Application) DNA crosslinking, DNA synthesis inhibition, nuclear division inhibition, apoptosis induction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.