Learn More
Novus Biologicals™ Mitomycin C Inhibitor

Description
Biological Activity:R-(-)-enantiomer of gossypol. Mimics the BH3 domains of Bcl-2, Bcl-XL and Mcl-1. Disrupts heterodimerization of Bcl-2 with proapoptotic family members. Induces apoptosis in vitro through activation of caspase-9; cytotoxic to multiple myeloma and drug-resistant cell lines. Delays onset of androgen-independent growth of VCaP prostate cancer xenografts in vivo.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 50-07-7 |
| Formulation | C15H18N4O5 |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 10 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 334.33g/mol |
| Solubility | Soluble to 5mM in water, 100mM in DMSO |
| Content And Storage | 4°C |
| Target | Mitomycin C |
| Color | Dark Purple |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.