Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mitomycin C, Solid, 99% Purity, 2 mg

Catalog No. 502584568
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4568 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4568 Supplier Abcam Supplier No. AB1207972MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent anticancer and antibiotic agent that crosslinks DNA to inhibit synthesis and induce apoptosis.

  • Molecular weight: 334.33 Da
  • Purity: >99%
  • Physical form: Solid
  • Key properties: Anticancer and antibiotic agent, crosslinks DNA, inhibits DNA synthesis and nuclear division, and induces apoptosis

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule DNA
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Buffer 99% Mitomycin C
Content And Storage Store at 4°C.
Form Solid
Formulation Constituents: 99% Mitomycin C
Quantity 2 mg
For Use With (Application) Anticancer agent, Antibiotic agent, DNA crosslinking, Inhibition of DNA synthesis, Inhibition of nuclear division, Induction of apoptosis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.