Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitomycin C, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139721
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139721 100 mg
S139805 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139721 Supplier G-Biosciences Supplier No. RC1056
Add to Cart
Add to Cart

Mitomycin C is an antineoplastic antibiotic isolated from Streptomyces caespitosus. It is effective against gram-negative and gram-positive bacteria and a variety of cancer cells, including Ehrlich ascites tumor cells.

Mitomycin C has been used to produce mitotically inactivated feeder cells and has been shown to increase recombination during fruit fly meiosis. It is capable of killing mutant strains of Arabidopsis that are defective for recombination during meiosis.

Chemical Identifiers

Molecular Weight (g/mol) 334.33 g/mol

Specifications

Boiling Point 581.8°C
Quantity 100 mg
Percent Purity 98%
Grade Laboratory
Physical Form Crystalline Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.