Learn More
Johannes Lieder Mitosis and Meiosis Set I
SCI_ED
Catalog No. S02817 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Composite
Catalog No. Mounting Type
S02817 Composite
1 options
Catalog No. S02817 Supplier Johannes Lieder Supplier No. 5150
Add to Cart
Add to Cart
5150 Mitosis and Meiosis Set I, 6 selected microscope slides
  • 5150 Mitosis and Meiosis Set I
  • 6 selected Microscope Slides
  • Mitosis, l.s. from Allium root tips showing all stages of plant mitosis carefully stained with iron
  • hematoxyline after Heidenhain
  • Mitotic stages in sec. through red bone marrow of mammal
  • Meiotic and mitotic stages in sec. of Salamandra testis. Many meiotic and mitotic stages can be observed
  • Lilium, anther t.s., microspore mother cells showing telophase of first and prophase of second (homeotypic) division
  • Giant chromosomes, smear from salivary gland of Chironomus, carefully fixed and stained
  • Ascaris megalocephala embryology. Sec. of uteri showing maturation stages (meiosis). Polar bodies can be seen.
Specifications
Classification Cell Biology
Material Elastin Stained Glass
Mounting Type Composite
Type Microscope Slides
Slide Type Prepared Slide