Learn More
Johannes Lieder Mitosis and Meiosis Set II
SCI_ED
Catalog No. S02816 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S02816 Cross and Longitudinal Section
1 options
Catalog No. S02816 Supplier Johannes Lieder Supplier No. 5170
Add to Cart
Add to Cart
5170 Mitosis and Meiosis Set II, 5 selected microscope slides
  • 5170 Mitosis and Meiosis Set II
  • 5 selected Microscope Slides
  • Mitosis, l.s. from Vicia faba (bean) root tips showing all mitotic stages. Iron hematoxyline stained
  • Lilium, anther t.s., microspore mother cells showing telophase of first and prophase of second (homeotypic) division
  • Mitotic stages in sec. of whitefish blastula showing spindles
  • Spermatogenesis with meiotic and mitotic stages, sec. of testis of Carausius, grasshopper, carefully stained
  • Paramaecium, in fission, nuclei stained
Specifications
Classification Cell Biology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Type Microscope Slides
Slide Type Prepared Slide