Learn More
Cayman Chemical MitoTEMPO (hydrate), 1569257-94-8, 10 mg
SDP
Supplier:  Cayman Chemical 1662110
Encompass_Preferred
Molecular Formula C29H35N2O2P • Cl [H2O], Purity ≥95%, Formula Weight 528.0; A mitochondria-targeted superoxide dismutase mimetic that possesses superoxide and alkyl radical scavenging properties.
Catalog No. NC1229394
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.