Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitoxantrone dihydrochloride, Tocris Bioscience™
SDP

Catalog No. 425050
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-505-0 Supplier R&D Systems Supplier No. 4250/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Topoisomerase II inhibitor; immunosuppressive and antineoplastic agent

Type II DNA topoisomerase inhibitor. Disrupts DNA synthesis and repair and induces damage by DNA cross-linking. Also inhibits PIM1 kinase (IC50 = 51 nM). Immunomodulatory, antineoplastic and chemotherapeutic agent. Also USP11 inhibitor (IC50= 3.15 μM). Induces cell death of pancreatic cancer cell lines expressing wild-type BRCA2.

Chemical Identifiers

CAS 70476-82-3
Molecular Formula C22H30Cl2N4O6
Molecular Weight (g/mol) 517.404
InChI Key ZAHQPTJLOCWVPG-UHFFFAOYSA-N
Synonym mitoxantrone hydrochloride, mitoxantrone dihydrochloride, mitoxantrone hcl, novantrone, novantron, immunex, bisantrone, unii-u6usw86rd0, dhad
PubChem CID 51082
ChEBI CHEBI:50727
IUPAC Name 1,4-dihydroxy-5,8-bis[2-(2-hydroxyethylamino)ethylamino]anthracene-9,10-dione;dihydrochloride
SMILES C1=CC(=C2C(=C1NCCNCCO)C(=O)C3=C(C=CC(=C3C2=O)O)O)NCCNCCO.Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 517.4
Percent Purity >98%
Chemical Name or Material Mitoxantrone dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.