Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Indicator Solution, Methyl Red-Methylene Blue, Ricca Chemical
SDP

Catalog No. 516416 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5164-16 500 mL Natural Poly Bottle
5164-4 120 mL Natural Poly Bottle
R51640001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5164-16 Supplier Ricca Chemical Company Supplier No. 516416
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Mixed Indicator Solution
CAS 64-17-5, 67-56-1, 845-10-3, 7220-79-3
Grade Indicator
Concentration or Composition (by Analyte or Components) Methyl Red - Methylene Blue
Packaging Natural Poly Bottle
Color Dark Green
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.