Description
Includes
Mounting board with silk-screened scale; eight wires of the same gauge but of different materials: silver, aluminum, copper, nicrome, iron, stainless steel, constantan; and detailed instructions with graphs
Students can calculate the resistivity of the higher resistance wires and compare them with theoretical values. By graphing the resitivity, they can see the linear nature of resistance in wires.
- Convenient scale is silk-screened onto the mounting board for easy measurements
- Banana plugs on either end allow for easy connections
Specifications
Specifications
| For Use With (Application) | To calculate the resistivity of the higher resistance wires and compare them with theoretical values |
| Includes | Mounting board with silk-screened scale, eight wires of the same gauge but of different materials: silver, aluminum, copper, nicrome, iron, stainless steel, constantan and detailed instructions with graphs |
| Product Type | Mixed Materials Resistance Board |