Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mixed Multi-Element Calibration Standard 4A, SPEX CertiPrep™
SDP

Catalog No. MXSTD4A100N Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MXSTD4A100N 125 mL
MXSTD4A500N 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MXSTD4A100N Supplier SPEX CertiPrep Supplier No. MXSTD4A100N
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Mixed Calibration Standards

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Mixed Calibration Std 4A, without Mercury
Packaging Triple-leached LDPE bottle
Name Note Without Mercury
CAS 7440-66-6, 7732-18-5, 7631-86-9, 7440-32-6, 7440-47-3
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al(100mg/L),SiO2(100mg/L),Ti(100mg/L)
Technique Calibration of Conductivity Instrument
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, SiO2, Ti
Solvent or Matrix Nitric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Grade Assurance
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.