Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
GREENER_CHOICE
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
$652.00 - $945.00
Specifications
Tubing Style TPE
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Port Detail Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Port Detail Price Quantity & Availability  
SH3066101
GREENER_CHOICE
View Documents
Thermo Scientific™
SH30661.01
50 L 4 x Top-mounted Ports and Tubing
Each for $698.00
Only null left
Add to Cart
SH3066102
GREENER_CHOICE
View Documents
Thermo Scientific™
SH30661.02
100 L 4 x Top-mounted Ports and Tubing
Each for $781.50
Only null left
Add to Cart
SH3066103
GREENER_CHOICE
View Documents
Thermo Scientific™
SH30661.03
200 L 4 x Top-mounted Ports and Tubing
Each for $945.00
Only null left
Add to Cart
SH3068701
GREENER_CHOICE
View Documents
Thermo Scientific™
SH30687.01
50 L Powder Port
Each for $652.00
Only null left
Add to Cart
Description
Description
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Specifications
Specifications
TPE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.