Learn More
Apexbio Technology LLC MJ33 (lithium salt)(Synonyms: MJ33 lithium salt, MJ33 Li salt, MJ33 inhibitor), 5mg, CAS: 1007476-63-2;199106-13-3.
SDP
Supplier:  Apexbio Technology LLC C53465
Encompass_Preferred
MJ33 (lithium salt CAS 1007476-63-2 199106-13-3) is a small molecule inhibitor of the acidic calcium-independent phospholipase A2 (aiPLA2) activity of peroxiredoxin-6 (Prdx6) Prdx6 exhibits dual enzymatic functions encompassing both glutathione peroxidase and PLA2 activities It plays a role in lung surfactant lipid metabolism particularly processing dipalmitoylphosphatidylcholine MJ33 selectively suppresses aiPLA2 activity as demonstrated through inhibition of phosphorylation-stimulated recombinant and native Prdx6 in vitro Moreover MJ33 has shown protective effects in animal models attenuating ROS generation inflammatory cytokine release NF- B activation and tissue damage in LPS-induced acute lung injury highlighting its utility for inflammation-related biomedical research
Catalog No. 50-000-61397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.