Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MK 0677
GREENER_CHOICE

Catalog No. p-7110798
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-721-0 10 mg
52-725-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-721-0 Supplier Tocris Bioscience™ Supplier No. 5272/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity ghrelin receptor agonist

MK 0677 is a high affinity ghrelin receptor agonist (pKi = 8.14). Growth hormone (GH) secretagog; stimulates GH release from rat pituitary cells in vitro (EC50 = 1.3 nM) and enhances GH plasma levels in vivo. Also attenuates isoproterenol -induced lipolysis in rat adipocytes in vitro. Orally bioavailable.

Specifications

Chemical Name or Material 2-Amino-N-[(1 R)-2-[1,2-dihydro-1-(methylsulfonyl)spiro[3 H-indole-3,4 '-piperidin]-1 '-yl]-2-oxo-1-[(phenylmethoxy)methyl]ethyl]-2-methylpropanamide methanesulfonate
CAS 159752-10-0
Synonym Ibutamoren mesylate, L-163,191
Quantity 10 mg
Target Ghrelin Receptor Agonists
Molecular Formula C27H36N4O5S.CH3SO3H
Formula Weight 624.77
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.