Learn More
Selleck Chemical LLC MK-0752-5mg
SDP
Supplier:  Selleck Chemical LLC S26605MG
Encompass_Preferred
MK-0752 is a moderately potent γ-secretase inhibitor, which reduces Aβ40 production with IC50 of 5 nM. Phase 1/2.
Catalog No. 50-238-9963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.