Learn More
Apexbio Technology LLC MK 0893, 5mg. Cas: 870823-12-4 MFCD: MFCD11616198. Glucagon receptor/IGF-1R antagonist
SDP
Supplier:  Apexbio Technology LLC A36085
Encompass_Preferred
MK 0893 is an inhibitor of both glucagon receptor and IGF-1R with IC50 values of 6.6nM and 6nM, respectively [1, 2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4814