Learn More
TARGETMOL CHEMICALS INC MK-6892 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ0108
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MK-6892 is a selective and full agonist for the high-affinity nicotinic acid receptor GPR109A (Ki 4 nM; GTP(gamma)S EC50 16 nM). purity: 99%
Catalog No. 50-257-8827
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.