Learn More
Selleck Chemical LLC MK-8719 5mg
SDP
Supplier:  Selleck Chemical LLC S88905MG
Encompass_Preferred
MK-8719 is a selective and potent small molecule inhibitor of the O-GlcNAcase (OGA) enzyme with ki of 7.9nM for hOGA. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3420
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.