Learn More
Selleck Chemical LLC MK-8745 10mg 885325-71-3
SDP
Supplier:  Selleck Chemical LLC S706510MG
Encompass_Preferred
MK-8745 is a potent and selective Aurora A inhibitor with IC50 of 0.6 nM, more than 450-fold selectivity for Aurora A over Aurora B. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.