Learn More
Apexbio Technology LLC MK-8776 (SCH-900776) 891494-63-6 10mg
SDP
Supplier:  Apexbio Technology LLC A847710
Encompass_Preferred
MK-8776 (SCH-900776 CAS number 891494-63-6) is a selective small molecule inhibitor targeting the serine/threonine kinase checkpoint kinase 1 (Chk1) Chk1 is involved in detecting DNA damage and regulating DNA replication particularly in stabilizing replication forks during genotoxic stress SCH-900776 selectively inhibits Chk1 activity (IC50 3 nM) over Chk2 and Cdk3 In cellular assays SCH-900776 treatment results in dose-dependent inhibition of Chk1 autophosphorylation at Ser296 accompanied by increased phosphorylation at Ser345 a cellular marker consistent with checkpoint disruption In preclinical models combining SCH-900776 with DNA-damaging agents such as gemcitabine enhances anti-tumor efficacy marking its potential utility in cancer research
Catalog No. 50-000-61717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.