Learn More
Enzo Life Sciences MKP-2 (human), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4620100
Encompass_Preferred
Alternative name: MAP kinase phosphatase 2, DUSP4, Dual specificity protein phosphatase 4, HVH2. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, 200mM NaCl, 5mM DTT, 0.1 % Tween-20, 10% glycerol. Source: Produced in Sf9 insect cells. Recombinant human MKP-2 fused at the N-terminus to a His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2036
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.