Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MKS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 139751AP150UL
Specifications
| MKS3 | |
| Polyclonal | |
| Unconjugated | |
| TMEM67 | |
| JBTS6, Meckel syndrome type 3 protein, MECKELIN, MKS3, TMEM67, TNEM67, transmembrane protein 67 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 329795, 91147 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5HYA8, Q8BR76 | |
| Tmem67 | |
| MKS3 Fusion Protein Ag5009 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction