Learn More
Apexbio Technology LLC ML-098 25mg
SDP
Supplier:  Apexbio Technology LLC C480725
Encompass_Preferred
ML-098 (CAS 878978-76-8) is a selective small-molecule activator targeting the GTP-binding protein Rab7 By effectively modulating Rab7 activity ML-098 facilitates intracellular membrane trafficking processes In vitro studies demonstrate that ML-098 activates Rab7 with an EC50 value of 77 6 nM highlighting its potency in regulating this GTPase s function ML-098 is commonly employed in biomedical research focused on cellular transport mechanisms and neurodegenerative disease pathogenesis
Catalog No. 50-000-62964
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.