Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 154

Catalog No. 516150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-615-0 50 mg
51-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-615-0 Supplier Tocris Bioscience™ Supplier No. 5161/50
This item is not returnable. View return policy

Potent neuropeptide S receptor (NPSR) antagonist

ML 154 is a potent neuropeptide S receptor (NPSR) antagonist (pA2 = 9.98). Inhibits neuropeptide S-induced ERK phosphorylation over cAMP responses and calcium responses (IC50 values are 9.3, 22.1 and 36.5 nM, respectively). Appears to modulate addictive behavior in vivo. Displays no activity against vasopressin V1B receptors. Brain penetrant.

Specifications

Purity >99%
Quantity 50 mg
Product Type ML 154
Molecular Weight (g/mol) 545.47
Inhibitors ML 154

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.