Learn More
Abcam ML-162, 1MG
Supplier:  Abcam AB2869991MG
Encompass_Preferred
MW 477.4 Da, Purity >=98%. ML-162 is a GPX4 (glutathione peroxidase 4) inhibitor that is selectively lethal to mutant RAS oncogene-expressing cell lines (IC₅₀s = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5878
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.