Learn More
Selleck Chemical LLC ML 210 2mg
SDP
Supplier:  Selleck Chemical LLC S07882MG
Encompass_Preferred
ML-210 (CID 49766530) is a selective covalent inhibitor of cellularglutathione peroxidase 4 (GPX4) with EC50 of 0.04 ?M and inducesferroptosis. ML-210 can selectively kill cells induced to express mutantRAS. ML-210 exhibits anti-cancer activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.