Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 348
SDP

Catalog No. 534550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-455-0 50 mg
53-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-455-0 Supplier Tocris Bioscience™ Supplier No. 5345/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective and reversible lysophospholipase 1 (LYPLA1) inhibitor

ML 348 is a selective and reversible lysophospholipase 1 (LYPLA1) inhibitor (IC50 = 210 nM). Exhibits 14-fold selectivity for LYPLA1 over LYPLA2. Also selective over a panel of ∽30 other serine hydrolases.

Specifications

Purity >99%
Quantity 50 mg
Product Type ML 348
Molecular Weight (g/mol) 415.79
Inhibitors ML 348

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.