Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam ML-9, Protein Kinase Inhibitor, >98% Purity, 10 mg

Catalog No. 502706081
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6081 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6081 Supplier Abcam Supplier No. AB14379710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broad-spectrum ATP-competitive protein kinase inhibitor and Ca²± channel blocker.

  • Molecular weight: 361.3 Da
  • Purity: >98%
  • Description: Broad spectrum ATP-competitive protein kinase inhibitor (Ki values are 32, 4, and 54 μM for PKA, MLCK, and PKC respectively)
  • Functional activity: Ca2+ channel blocker; shows vasorelaxant effects in vivo
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% ML-9
Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule PKA, MLCK, PKC, Ca²± Channel
Quantity 10 mg
For Use With (Application) Protein kinase inhibition, Ca²± channel blockade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.