Learn More
Abcam ML-9, Protein Kinase Inhibitor, >98% Purity, 250 mg
Description
- Molecular weight: 361.3 Da
- Purity: >98%
- Mechanism: Broad spectrum ATP-competitive protein kinase inhibitor
- Ki values: 32 μM (PKA), 4 μM (MLCK), 54 μM (PKC)
- Function: Ca²+ channel blocker
- Biological effect: Shows vasorelaxant effects in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Protein Kinase (PKA, MLCK, PKC), Ca²± Channel |
| Buffer | 100% ML-9 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% ML-9 |
| Quantity | 250 mg |
| For Use With (Application) | Protein kinase inhibition, Ca²± channel blockade, vasorelaxation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.