Learn More
Apexbio Technology LLC ML133 HCl 1222781-70-5 50mg
SDP
Supplier:  Apexbio Technology LLC B219950
Encompass_Preferred
ML133 HCl (CAS 1222781-70-5) is a small-molecule inhibitor that selectively targets the inward rectifier potassium channel K sub ir /sub 2 1 It exhibits concentration-dependent inhibition with an IC sub 50 /sub of 1 8 M at pH 7 4 and 290 nM at pH 8 5 The compound demonstrates minimal activity against K sub ir /sub 4 1 and K sub ir /sub 7 1 and does not affect K sub ir /sub 1 1 channels ML133 HCl is utilized in biomedical research to investigate the physiological and pathological roles of K sub ir /sub 2 1 supporting studies on potassium channel function cardiac electrophysiology and related ion channel disorders
Catalog No. 50-000-89691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.