Learn More
Apexbio Technology LLC ML241 1346528-06-0 25mg
SDP
Supplier:  Apexbio Technology LLC B610725
Encompass_Preferred
ML241 (CAS 1346528-06-0) is a potent and selective inhibitor of the p97 ATPase (also known as valosin-containing protein VCP) an AAA family ATPase involved in diverse cellular processes including degradation of misfolded proteins membrane fusion protein aggregate regulation and autophagosome maturation ML241 exhibits an in vitro IC50 of approximately 100 nM for p97 inhibition In cellular models ML241 impedes the degradation of p97-dependent but not p97-independent proteasomal substrates and disrupts the endoplasmic reticulum-associated degradation (ERAD) pathway Currently ML241 is utilized in preclinical research to investigate p97-related cellular mechanisms in cancer and neurodegenerative disease contexts
Catalog No. 50-000-91988
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.